• USA Home
  • H4150 - 15β-Hydroxytestosterone

EMAIL THIS PAGE TO A FRIEND
H4150 Sigma

15β-Hydroxytestosterone

~50 μg/mL in methanol, ≥98% (HPLC)

Synonym: 4-Androstene-15β,17β-diol-3-one

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Biomarkers and Metabolomics Standards, Hormone Standards
InChI Key   KYGUQDTWUBBBSD-XJBBOAPUSA-N
assay   ≥98% (HPLC)
drug control   USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   ~50 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1230 3/PG 2
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?