| Related Categories | 3-D Stem Cell Culture, 3D Matricies for Stem Cell Biology, Attachment Factors, Cell Biology, Cell Culture, |
| biological source | from synthetic |
| storage temp. | −20°C |
Live Chat and Frequently Asked Questions
are available for this Product.
A synthetic peptide nanofiber scaffold, HydroMatrix offers the precision and control of a synthesized matrix with the natural three dimensional architecture of highly crosslinked peptide hydrogel. The HydroMatrix scaffold self-assembles from fluid precursors into a highly cross-linked peptide 3-dimensional hydrogel in response to changes in temperature or ionic strength. By adjusting the concentration of the HydroMatrix solution, researchers are able to control the flexibility of the 3-D architecture, and tailor the structure to meet their individual needs.
HydroMatrix promotes cell growth and migration and has been shown to support the proliferation of many cell types, including neural stem cells, neurons, glia, astrocytes, fibroblasts, and keratinocytes.
HydroMatrix is a trademark of Sigma-Aldrich Co. LLC
Customers Also Viewed
liquid, BioReagent, suitable for cell culture
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?