• USA Home
  • H4252 - m-Hydroxyphenylacetic acid

EMAIL THIS PAGE TO A FRIEND
H4252 Sigma

m-Hydroxyphenylacetic acid

Synonym: 3-Hydroxyphenylacetic acid

Purchase

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

H49901 3-Hydroxyphenylacetic acid, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?