• USA Home
  • H4506 - Hemolymph plasma powder from Limulus polyphemus

EMAIL THIS PAGE TO A FRIEND
H4506 Sigma

Hemolymph plasma powder from Limulus polyphemus

lyophilized powder

Purchase

Properties

Related Categories Biochemicals and Reagents, Coagulation Proteins and Reagents, Plasma & Blood Proteins, Proteins and Derivatives
form   lyophilized powder
composition   Protein, ~60% biuret
storage temp.   2-8°C

Description

Other Notes

Prepared from Horseshoe crab blood after the Amoebocyte lysate (E-Toxate®) has been removed.

Quality

Crude

Physical form

Lyophilized powder, sodium chloride and other naturally occurring components, including lectins

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?