• USA Home
  • H4524 - Histone from calf thymus

EMAIL THIS PAGE TO A FRIEND
H4524 Sigma

Histone from calf thymus

suitable for substrate for protein kinase C

Purchase

Description

Application

Commonly used for chromosome-reconstitution studies.

Biochem/physiol Actions

Suitable substrate for protein kinase C.

Features and Benefits

Do not confuse Sigma "Type" designations with the "Fraction" designations used by Luck and co-workers.

Other Notes

Characterized as mainly subgroup f1 by SDS-PAGE

Preparation Note

Isolated as a lysine-rich fraction.

Suitability

Phosphorylation of this histone may also be suitable in other protein kinase systems.

Price and Availability


Amplified Detection

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?