• USA Home
  • H4527 - 15-Hydroxypentadecanoic acid

EMAIL THIS PAGE TO A FRIEND
H4527 Sigma

15-Hydroxypentadecanoic acid

≥99% (capillary GC)

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, C13 to C42+, Carbonyl Compounds, Carboxylic Acids,
InChI Key   BZUNJUAMQZRJIP-UHFFFAOYSA-N
assay   ≥99% (capillary GC)
mp   85-89 °C(lit.)
storage temp.   2-8°C

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?