EMAIL THIS PAGE TO A FRIEND
H4640 Sigma

HSV Peptide

≥97% (HPLC), lyophilized powder

Synonym: Herpes simplex virus human

Purchase

Properties

Related Categories HSV, Molecular Biology, Proteomics, Purification and Detection, Recombinant Protein Expression and Analysis More...
assay   ≥97% (HPLC)
form   lyophilized powder
mol wt   mol wt 1367.4 Da
storage temp.   −20°C

Description

Amino Acid Sequence

Lys-Gln-Pro-Glu-Leu-Ala-Pro-Glu-Asp-Pro-Glu-Asp

Application

Competitive Inhibition of antibody binding by HSV antibodies

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?