• USA Home
  • H4648 - 1-O-Palmitol-2-(N-methylcarbamyl)-sn-glycero-3-phosphocholine

EMAIL THIS PAGE TO A FRIEND
H4648 Sigma

1-O-Palmitol-2-(N-methylcarbamyl)-sn-glycero-3-phosphocholine

≥98% (TLC), white powder

Synonym: mcPAF(C16), mcPlatelet activating factor

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, Lipids in Cell Signaling,
InChI Key   FNFHZBKBDFRYHS-RUZDIDTESA-N
assay   ≥98% (TLC)
form   white powder
solubility   H2O: soluble20 mg/mL
storage temp.   −20°C

Description

Biochem/physiol Actions

Nonhydrolyzable form of PAF[C16]. Activates platelets and exhibits potent inflammatory activity, mediator of cellular damage due to cerebral ischemia.

Other Notes

hygroscopic

Features and Benefits

This compound is featured on the PAF Receptor page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?