• USA Home
  • H4659 - Anti-Heme Oxygenase-2 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
H4659 Sigma

Anti-Heme Oxygenase-2 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-HO-2

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, H1-HE, Primary Antibodies
species reactivity   mouse, rat
application(s)   western blot (chemiluminescent): 0.5-2 μg/mL using mouse or rat brain extracts
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt 34 kDa
shipped in   wet ice
storage temp.   2-8°C
Gene Information   human ... HMOX2(3163)
mouse ... Hmox2(15369)
rat ... Hmox2(79239)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 247-258 of the rat heme oxygenase-2 protein conjugated to KLH. This sequence is the same in mouse HO-2, and contains only one amino acid substitution with human HO-2.

Physical form

Solution in 0.05 M sodium phosphate buffer, pH 7.5, containing 0.2% gelatin and 0.1% sodium azide

Specificity

By immunoblotting, the antibody detects recombinant heme oxygenase-2 (34 kDa). It is not recommended for immunohistochemistry.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Personal Protective Equipment  dust mask type N95 (US), Eyeshields, Gloves
RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?