EMAIL THIS PAGE TO A FRIEND
H5036 Sigma
Bottle holders
single, diam. 3 3/4 in. , hole
Purchase
Properties
diam.   3 3/4 in. , hole
Description
General description
Keeps squeeze and other bottles from tipping over as bottle becomes top heavy. Units feature 3 1/8 in. or 3 3/4 in. ID holes and mount instantly with 3-M double-faced tape. Can be screw-mounted on wall, hood or secured to benchtop.
Saves on solvents and prevents spills. Useful next to sinks or other solvent wash areas.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
H5161 Bottle holders, double, diam. 3 3/4 in. , hole
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?