• USA Home
  • H5152 - Heparin sodium salt solution from porcine intestinal mucosa

EMAIL THIS PAGE TO A FRIEND
H5152 Sigma

Heparin sodium salt solution from porcine intestinal mucosa

5 μg/mL in H2O (no preservative), Tyramine and FITC labeled, average mol wt 3,600-7,500

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Polysaccharide More...
description   low molecular weight
anti IIa activity   ~3 U/mg
secondary activity   60-90 U/mg anti-Xa
mol wt   average mol wt 3,600-7,500
concentration   5 μg/mL in H2O (no preservative)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?