• USA Home
  • H5398 - S-(2-Hydroxy-5-nitrobenzyl)-6-thioguanosine

EMAIL THIS PAGE TO A FRIEND
H5398 Sigma

S-(2-Hydroxy-5-nitrobenzyl)-6-thioguanosine

≥90% (TLC)

Synonym: 2-Amino-6-[(2-hydroxy-5-nitrobenzyl)thio]-9-β-D-ribofuranosylpurine

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   JXIKYSYYTDTHBG-SDPXSBNPSA-N
assay   ≥90% (TLC)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?