EMAIL THIS PAGE TO A FRIEND
H5517 Sigma

2-Hydroxyestriol

≥90% (HPLC), powder

Synonym: 1,3,5(10)-Estratriene-2,3,16α,17β-tetrol, 2,3,16α,17β-Tetrahydroxy-1,3,5(10)-estratriene

  • CAS Number 1232-80-0

  • Empirical Formula (Hill Notation) C18H24O4

  • Molecular Weight 304.38

  •  MDL number MFCD00056550

  •  PubChem Substance ID 24895695

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, Cytokines, Growth Factors and Hormones,
InChI Key   ZUGCDOZAVASBQT-SLVREZFOSA-N
assay   ≥90% (HPLC)
form   powder
color   off-white
storage temp.   −20°C

Price and Availability


Better Together

Discover your Target

All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?