EMAIL THIS PAGE TO A FRIEND
H5536 Sigma

Humanin

solid

  • Empirical Formula (Hill Notation) C119H203N34O32S2

  • Molecular Weight 2686.22

  •  MDL number MFCD03787970

Purchase

Description

Amino Acid Sequence

Met-Ala-Pro-Arg-Gly-Phe-Ser-Cys-Leu-Leu-Leu-Leu-Thr-Ser-Glu-Ile-Asp-Leu-Pro-Val-Lys-Arg-Arg-Ala

Biochem/physiol Actions

Suppresses neuronal cell death in cells expressing amyloid precursor protein mutants related to familial Alzheimer′s disease.

Price and Availability


Amplified Detection

Making Headway

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?