• USA Home
  • H5631 - 19-Hydroxycholesterol 3-acetate

EMAIL THIS PAGE TO A FRIEND
H5631 Sigma

19-Hydroxycholesterol 3-acetate

Synonym: 5-Cholestene-3β,19-diol 3-acetate

  • CAS Number 750-59-4

  • Empirical Formula (Hill Notation) C29H48O3

  • Molecular Weight 444.69

  •  MDL number MFCD00201432

  •  PubChem Substance ID 329815220

Purchase

Properties

InChI Key   ZUYKAEVLUWUNMD-KVHJHAPGSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?