• USA Home
  • H5760 - Hydrocortisone 21-acetate

EMAIL THIS PAGE TO A FRIEND
H5760 Sigma

Hydrocortisone 21-acetate

Sigma Reference Standard

Synonym: 11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-acetate, 17-α-Hydroxycorticosterone acetate, 17-Hydroxycorticosterone 21-acetate, 21-Acetoxy-4-pregnene-11β,17α-diol-3,20-dione, 4-Pregnene-11β,17α,21-triol-3,20-dione 21-acetate

  • CAS Number 50-03-3

  • Empirical Formula (Hill Notation) C23H32O6

  • Molecular Weight 404.50

  •  Beilstein Registry Number 2066841

  •  EC Number 200-004-4

  •  MDL number MFCD00037714

Purchase

Properties

grade   Sigma Reference Standard
InChI Key   ALEXXDVDDISNDU-JZYPGELDSA-N
packaging   vial of 250 mg
mp   223 °C (dec.)(lit.)

Description

Packaging

Supplied in amber screw-cap vials

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
GM8960000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

H4126 Hydrocortisone 21-acetate, ≥98% (HPLC)
H3035 Hydrocortisone 21-acetate, meets USP testing specifications

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?