• USA Home
  • H5785 - HIV Protease Substrate VIII

EMAIL THIS PAGE TO A FRIEND
H5785 Sigma

HIV Protease Substrate VIII

>96% (HPLC)

  • Empirical Formula (Hill Notation) C42H66N10O13

  • Molecular Weight 919.03

  •  MDL number MFCD03444583

Purchase

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?