• USA Home
  • H6005 - Histone from calf thymus

EMAIL THIS PAGE TO A FRIEND
H6005 Sigma

Histone from calf thymus

Type II-S

Purchase

Properties

Related Categories Biochemicals and Reagents, Bovine, Proteins and Derivatives More...
type   Type II-S
storage temp.   2-8°C

Description

Features and Benefits

Do not confuse Sigma "Type" designations with the "Fraction" designations used by Luck and co-workers.

Preparation Note

Prepared by extraction in 1M NaCl solution, precipitation in water, acid extraction, and reprecipitation with alcohol.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?