• USA Home
  • H6013 - (S,S)-(−)-Hydrobenzoin

EMAIL THIS PAGE TO A FRIEND
H6013 Sigma

(S,S)-(−)-Hydrobenzoin

Synonym: (S,S)-(−)-1,2-Diphenyl-1,2-ethanediol

Purchase

Properties

storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?