• USA Home
  • H6035 - Anthranilyl-HIV Protease Substrate V

EMAIL THIS PAGE TO A FRIEND
H6035 Sigma

Anthranilyl-HIV Protease Substrate V

>95% (HPLC)

Synonym: 2-Aminobenzoyl-Ala-Arg-Val-Nle-p-nitro-Phe-Glu-Ala-Nle-NH2

  • Empirical Formula (Hill Notation) C50H76N14O13

  • Molecular Weight 1081.22

  •  MDL number MFCD03453612

Purchase

Price and Availability


All labs need water

Biomedical Applications

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?