• USA Home
  • H6141 - 19-Hydroxydeoxycorticosterone 21-acetate

EMAIL THIS PAGE TO A FRIEND
H6141 Sigma

19-Hydroxydeoxycorticosterone 21-acetate

~90%

Synonym: 21-Acetoxy-19-hydroxy-4-pregnene-3,20-dione, 4-Pregnene-19,21-diol-3,20-dione 21-acetate

Purchase

Properties

assay   ~90%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?