• USA Home
  • H6259 - Hydrocortisone 21-caprylate

EMAIL THIS PAGE TO A FRIEND
H6259 Sigma

Hydrocortisone 21-caprylate

Synonym: 11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-caprylate, 4-Pregnene-11β,17α,21-triol-3,20-dione 21-caprylate, Cortisol octanoate, Hydrocortisone 21-octanoate

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Corticosteroid, Cytokines, Growth Factors and Hormones, Hormones,
InChI Key   VWVPRYMOFYIOOZ-KAQKJVHQSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?