• USA Home
  • H6379 - Heptadecanoyl chloride

EMAIL THIS PAGE TO A FRIEND
H6379 Sigma

Heptadecanoyl chloride

~99%, liquid

Synonym: Margaroyl chloride

  • CAS Number 40480-10-2

  • Linear Formula CH3(CH2)15COCl

  • Molecular Weight 288.90

  •  Beilstein Registry Number 1781006

  •  MDL number MFCD00000743

  •  PubChem Substance ID 329815231

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   ICDQUAGMQCUEMY-UHFFFAOYSA-N
assay   ~99%
form   liquid
refractive index   n20/D 1.453(lit.)
bp   176 °C/4 mmHg(lit.)
density   0.883 g/mL at 25 °C(lit.)
storage temp.   −20°C

Description

Packaging

Sealed ampule.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

306657 Heptadecanoyl chloride, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?