• USA Home
  • H6387 - His-Cys-Lys-Phe-Trp-Trp

EMAIL THIS PAGE TO A FRIEND
H6387 Sigma

His-Cys-Lys-Phe-Trp-Trp

≥95% (HPLC)

Purchase

Description

Amino Acid Sequence

His-Cys-Lys-Phe-Trp-Trp

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Biochem/physiol Actions

Inhibits HIV integrase (IN)-mediated 3′-processing and integration of HIV DNA. Also active on integrases from HIV-2, feline immunodeficiency virus, and Moloney murine leukemia virus.

Price and Availability


Better Together

Discover your Target

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?