EMAIL THIS PAGE TO A FRIEND
H6397 Sigma
Hand shield, acrylic
for Pipetman pipettors
Purchase
Description
General description
Shields of clear 3/8 in. acrylic protect hand and forearm while pipetting most β-emitters, including 32P.
Shields are custom-designed to fit two popular pipetter styles; attaches using either the pipetter nose cone nut or an integral set screw.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?