• USA Home
  • H6529 - Sodium hippurate hydrate

EMAIL THIS PAGE TO A FRIEND
H6529 Sigma

Sodium hippurate hydrate

BioXtra, ≥99%

Synonym: Benzoylaminoacetic acid sodium salt, Hippuric acid sodium salt hydrate, N-Benzoylglycine sodium salt

  • CAS Number 532-94-5

  • Linear Formula C6H5CONHCH2COONa · xH2O

  • Molecular Weight 201.15 (anhydrous basis)

  •  Beilstein Registry Number 3638470

  •  EC Number 208-548-4

  •  MDL number MFCD00149113

  • Popular Documents:  RAMAN FTIR (PDF)  

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, I - Z, Modified Amino Acids
InChI Key   IOZRWVOSUXHESF-UHFFFAOYSA-M
product line   BioXtra
assay   ≥99%
impurities   ≤0.0005% Phosphorus (P)
  ≤0.1% Insoluble matter
solubility   H2O: soluble0.5 M at 20 °C, clear, colorless
anion traces   chloride (Cl-): ≤0.05%
  sulfate (SO42-): ≤0.05%
cation traces   Al: ≤0.0005%
  Ca: ≤0.0005%
  Cu: ≤0.0005%
  Fe: ≤0.0005%
  K: ≤0.005%
  Mg: ≤0.0005%
  NH4+: ≤0.05%
  Pb: ≤0.001%
  Zn: ≤0.0005%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?