• USA Home
  • H6658 - Heat Shock Protein 90 from Saccharomyces cerevisiae

EMAIL THIS PAGE TO A FRIEND
H6658 Sigma

Heat Shock Protein 90 from Saccharomyces cerevisiae

≥85% (SDS-PAGE), recombinant, expressed in Saccharomyces cerevisiae (haploid), lyophilized powder

Synonym: HSP 90

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Cell Signaling Enzymes, Cell Stress and Refolding, Enzymes, Inhibitors, and Substrates More...
recombinant   expressed in Saccharomyces cerevisiae (haploid)
assay   ≥85% (SDS-PAGE)
form   lyophilized powder
storage temp.   −20°C

Description

Physical form

Lyophilized from 20 mM MOPS, pH 7.4, 1 mM EDTA

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?