• USA Home
  • H6784 - HyPep® 4601 Protein Hydrolysate from wheat gluten

EMAIL THIS PAGE TO A FRIEND
H6784 Sigma

HyPep® 4601 Protein Hydrolysate from wheat gluten

Animal-component free, powder, suitable for cell culture

Synonym: Peptone from wheat gluten

Purchase

Properties

Related Categories CHO Platform, Cell Culture, Miscellaneous Reagents and Supplements, Reagents and Supplements, Serum-free Media,
Quality Level   GMP
form   powder
suitability   suitable for cell culture
storage temp.   2-8°C

Description

Other Notes

Animal-component free

Application

Recommended for use as a rich source of L-glutamine for medium supplementation at 1 to 5 g/L of medium. It also provides amino acids that support the general nutritional requirements of cells in culture.

Legal Information

HyPep is a registered trademark of Kerry Group

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles

Related Content

L-Glutamine in Cell Culture

Importance and uses of glutamine in serum-free eucaryotic, including hybridoma and Chinese Hamster Ovary (CHO) cell, cultures
Keywords: Alternative energy, Cell culture, Culture media, Deaminations, Metabolism, Oxidations, Transaminations, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?