• USA Home
  • H7023 - 1-(4-Hydroxyphenyl)-1H-tetrazole-5-thiol

EMAIL THIS PAGE TO A FRIEND
H7023 Sigma

1-(4-Hydroxyphenyl)-1H-tetrazole-5-thiol

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Tetrazoles
InChI Key   MOXZSKYLLSPATM-UHFFFAOYSA-N
mp   170 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

371599 1-(4-Hydroxyphenyl)-1H-tetrazole-5-thiol, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?