• USA Home
  • H7259 - trans-4-Hydroxy-L-proline β-naphthylamide

EMAIL THIS PAGE TO A FRIEND
H7259 Sigma

trans-4-Hydroxy-L-proline β-naphthylamide

Synonym: HydroxyPro-betaNA

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   RPDYJTXWIGUABB-KGLIPLIRSA-N
storage temp.   −20°C

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Application

Trans-4-Hydroxy-L-proline β-naphthylamide (HydroxyPro-βNA) is used for colorimetric determination of bacterial peptidase(s).

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?