• USA Home
  • H7270 - HC toxin from Helminthosporium carbonum

EMAIL THIS PAGE TO A FRIEND
H7270 Sigma

HC toxin from Helminthosporium carbonum

lyophilized powder

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, H,
InChI Key   GNYCTMYOHGBSBI-UHFFFAOYSA-N
assay   90-95% (TLC)
form   lyophilized powder
storage temp.   2-8°C

Description

Biochem/physiol Actions

Cyclic tetrapeptide fungal toxin selectively toxic to plants with susceptible host genotype.

Packaging

1, 5 mg in glass bottle

Price and Availability


Better Together

Discover your Target

All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?