• USA Home
  • H7285 - Hepatitis C virus NS4 antigen−Biotin

EMAIL THIS PAGE TO A FRIEND
H7285 Sigma

Hepatitis C virus NS4 antigen−Biotin

~1 mg/mL, ≥95% (SDS-PAGE), recombinant, expressed in E. coli

Purchase

Properties

Related Categories AIDS and Viral Research Reagents, Cell Biology, Cell Signaling and Neuroscience, Immune Cell Signaling and Blood, Immune System Regulation More...
recombinant   expressed in E. coli
assay   ≥95% (SDS-PAGE)
mol wt   mol wt 19 kDa (plus 114 kDa β-Gal tag)
concentration   ~1 mg/mL
storage temp.   −20°C
Gene Information   hepatitis C virus ... HCVgp1(951475)

Description

Biochem/physiol Actions

Positive control for HCV antibodies.

Other Notes

HCV NS4a+b [1658-1863]-Galactosidase-tagged, biotin conjugate.

Physical form

Solution in 8 M Urea, 20 mM Tris-HCl, pH 8.0, 10 mM 2-Mercaptoethanol.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?