EMAIL THIS PAGE TO A FRIEND
H7380 Sigma

Hirudin from leeches

lyophilized powder, 300-1,500 ATU/mg protein (ATU = antithrombin units)

Purchase

Description

Analysis Note

Protein determined by Lowry.

General description

Hirudin is a 7 kDa acidic protein containing 65 amino acid residues that has an isoelectric point of 3.5-4.0. It is not glycosylated and lacks tryptophan, arginine and methionine residues. It is chemically stable between pH 2-12 and at temperatures up to 80°C.

Unit Definition

One unit (ATU) will neutralize one NIH unit of thrombin at 37 °C, based on direct comparison to an NIH thrombin reference standard.

Physical form

Lyophilized from 0.045 M NaCl and 0.02 M Tris buffer, pH 7.5

Biochem/physiol Actions

The anticoagulant, hirudin, is the most potent natural inhibitor of both soluble and clot-bound thrombin. It forms a high-affinity 1:1 complex with thrombin, occluding both the proteolytic site and exosite I (fibrinogen and PAR recognition site). Hirudin blocks thrombus growth and platelet activation, and has been suggested as the clinically preferred anticoagulant (over heparin and citrate) for its specific mode of action and absence of side effects. It is not metabolized in the bloodstream of humans and is eliminated unchanged via kidney filtration.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
MR8440000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Plasma Derived Proteins and Enzymes

Despite their complexity, blood and plasma are abundant biological resources for the discovery of drug targets and biomarkers for human disease. It is estimated that plasma may contain as many as 40,...
BioFiles 2006, 1.5, 2.
Keywords: Applications, Gas chromatography, Type

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?