• USA Home
  • H7649 - Hemicellulase from Aspergillus niger

EMAIL THIS PAGE TO A FRIEND
H7649 Sigma

Hemicellulase from Aspergillus niger

soluble powder, ≥5 units/mg solid

Purchase

Properties

form   soluble powder
storage temp.   2-8°C

Description

Other Notes

View more information on enzymes for complex carbohydrate analysis at www.sigma-aldrich.com/enzymeexplorer

Quality

Also contains cellulase activity.

Unit Definition

One unit will liberate 1.0 μmole of D-galactose from hemicellulose per hr at pH 5.5 at 37°C (2 hour assay).

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

H2125 Hemicellulase from Aspergillus niger, powder, 0.3-3.0 unit/mg solid (using a β-galactose dehydrogenase system and locust bean gum as substrate)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?