• USA Home
  • H7755 - Histone from calf thymus

EMAIL THIS PAGE TO A FRIEND
H7755 Sigma

Histone from calf thymus

Type II-AS

Purchase

Properties

Related Categories Biochemicals and Reagents, Bovine, Proteins and Derivatives More...
type   Type II-AS
storage temp.   2-8°C

Description

Features and Benefits

Do not confuse Sigma "Type" designations with the "Fraction" designations used by Luck and co-workers.

Preparation Note

Prepared by extraction in 1 M NaCl solution, precipitation in water, acid extraction, dialysis and lyophilization.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?