• USA Home
  • H7762 - (±)-5-Hydroxy-(6E,8Z,11Z,14Z)-eicosatetraenoic acid methyl ester

EMAIL THIS PAGE TO A FRIEND
H7762 Sigma

(±)-5-Hydroxy-(6E,8Z,11Z,14Z)-eicosatetraenoic acid methyl ester

~100 μg/mL in methanol: water (65:35), ~98%

Synonym: 5-HETE methyl ester

Purchase

Properties

assay   ~98%
concentration   ~100 μg/mL in methanol: water (65:35)
shipped in   dry ice
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1230 3/PG 2
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?