• USA Home
  • H7880 - Hydroxymandelonitrile lyase from Sorghum Seedlings

EMAIL THIS PAGE TO A FRIEND
H7880 Sigma

Hydroxymandelonitrile lyase from Sorghum Seedlings

lyophilized powder, 2-4 units/mg protein

Synonym: 4-Hydroxymandelonitrile hydroxybenzaldehyde-lyase, Hydroxynitrile lyase

Purchase

Properties

Related Categories 4.1.x.x C-C bonds, 4.x.x.x Lyases, Biochemicals and Reagents, Enzyme Class Index, Enzymes, Inhibitors, and Substrates More...
form   lyophilized powder
composition   Protein, ~50% modified Warburg-Christian
storage temp.   −20°C

Description

Unit Definition

One unit will form 1.0 μmole of 4-hydroxybenzaldehyde and HCN from 4-hydroxymandelonitrile per minute at pH 5.4 at 25°C.

Physical form

Lyophilized powder containing buffer salts as phosphate and citrate

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?