• USA Home
  • H8153 - 7-Hydroxycoumarin-3-carboxylic acid ethyl ester

EMAIL THIS PAGE TO A FRIEND
H8153 Sigma

7-Hydroxycoumarin-3-carboxylic acid ethyl ester

~98%

Synonym: Ethyl umbelliferone-3-carboxylate

  • CAS Number 6093-71-6

  • Empirical Formula (Hill Notation) C12H10O5

  • Molecular Weight 234.20

  •  MDL number MFCD00017641

  •  PubChem Substance ID 24895820

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Fluorogenic More...
InChI Key   IETDBZQIWIJQJG-UHFFFAOYSA-N
assay   ~98%
storage temp.   −20°C
Gene Information   human ... MIF(4282)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?