• USA Home
  • H8377 - 5-Hydroxy-DL-tryptophan ethyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
H8377 Sigma

5-Hydroxy-DL-tryptophan ethyl ester hydrochloride

  • CAS Number 103404-89-3

  • Empirical Formula (Hill Notation) C13H16N2O3 · HCl

  • Molecular Weight 284.74

  •  MDL number MFCD00070408

  •  PubChem Substance ID 24895822

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   ZPSOVZKVIHRWEL-UHFFFAOYSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?