• USA Home
  • H8511 - Hydroxyalkoxypropyl-Dextran

EMAIL THIS PAGE TO A FRIEND
H8511 Sigma

Hydroxyalkoxypropyl-Dextran

Type X

Purchase

Properties

type   Type X
storage temp.   2-8°C

Description

Application

Lipophilic, hydrophobic gel for liquid chromatography.

Other Notes

Hydroxypropyl beaded dextran (Lipophilic Sephadex, LH-20-100), substituted with long chain alkyl ethers.

Substituted approx. 50% by weight with alkyl chains of length C18

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

H6258 Hydroxyalkoxypropyl-Dextran, Type VI

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?