• USA Home
  • H8523 - 6-Hydroxydopamine hydrobromide

EMAIL THIS PAGE TO A FRIEND
H8523 Sigma

6-Hydroxydopamine hydrobromide

≥90% (HPLC), solid

Synonym: 2,4,5-Trihydroxyphenethylamine hydrobromide, 2,5-Dihydroxytyramine hydrobromide, 2-(2,4,5-Trihydroxyphenyl)ethylamine hydrobromide, 6-OHDA

  • CAS Number 636-00-0

  • Linear Formula (HO)3C6H2CH2CH2NH2 · HBr

  • Molecular Weight 250.09

  •  Beilstein Registry Number 3713280

  •  EC Number 211-247-0

  •  MDL number MFCD00012894

  •  PubChem Substance ID 24895826

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

Related Categories Adrenergics, Cell Biology, Cell Signaling and Neuroscience, Neuroscience, Neurotransmission,
InChI Key   MLACDGUOKDOLGC-UHFFFAOYSA-N
assay   ≥90% (HPLC)
form   solid
color   tan to brown
mp   216-220 °C(lit.)
storage temp.   −20°C

Description

Biochem/physiol Actions

Neurotoxin that destroys catecholaminergic terminals.

Caution

Hygroscopic, light sensitive

Reconstitution

Dissolve in oxygen-free water containing 0.1% sodium metabisulfite or other antioxidants.

General description

Solutions should be freshly prepared and protected from exposure to light.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DC4600000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

H116 6-Hydroxydopamine hydrobromide, contains ascorbic acid as stabilizer, ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?