• USA Home
  • H8649 - Human Genetic Disease Analysis: A Practical Approach, 2nd ed.
EMAIL THIS PAGE TO A FRIEND
H8649 Sigma
Human Genetic Disease Analysis: A Practical Approach, 2nd ed.
  •  ISBN-10 0-19-963308-8
  •  ISBN-13 978-0-19-963308-1
Purchase
Properties
Related Categories Books and Software, Genetics Books, Labware More...
publication info   K.E. Davies, IRL Press at Oxford University Press, UK, 1993, 124 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
This new edition covers all the important methods now commonly in use, including synthetic oligonucleotide hybridization, pulsed-field gel electrophoresis, and fluorescence in situ hybridization. This book describes not only the analysis of gene mutations but also the functional organization of genes and chromosome regions.
Table of Contents
1. Fetal DNA Analysis, J.M. Old
2. The Use of Synthetic Oligonucleotides as Specific Hybridization Probes in the Diagnosis of Genetic Disorders, S.W. Thein, A. Ehsani, and R.B. Wallace
3. Pulsed-field Gel Electrophoresis in the Analysis of Genomic DNA and YAC Clones, J.T. den Dunnen, P.M. Grootscholten, and G.J.B. van Ommen
4. Fluorescent in situ Hybridization, V.J. Buckle and K.A. Rack
5. Fine Mapping of Genes: The Characterization of the Transcriptional Unit, M. Antoniou, E. deBoer, and F. Grosveld
6. Chromosome Analysis and Sorting by Flow Cytometry, S. Monard, L. Kearney, and B.D. Young
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?