• USA Home
  • H8655 - 17α-Hydroxy-11-deoxycorticosterone-[1,2-3H(N)]

EMAIL THIS PAGE TO A FRIEND
H8655 Sigma

17α-Hydroxy-11-deoxycorticosterone-[1,2-3H(N)]

≥95% (Radiochemical Purity), ethanol solution

Purchase

Properties

assay   ≥95% (Radiochemical Purity)
form   ethanol solution
extent of labeling   40-60 Ci/mmol
storage temp.   −20°C

Description

Packaging

serum bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?