• USA Home
  • H8771 - 1-O-Palmityl-2-acetyl-sn-glycero-3-phospho-(N,N,N-trimethyl)hexanolamine

EMAIL THIS PAGE TO A FRIEND
H8771 Sigma

1-O-Palmityl-2-acetyl-sn-glycero-3-phospho-(N,N,N-trimethyl)hexanolamine

~95%

Synonym: L-α-Phosphatidyl[N,N,N-trimethyl]hexanolamine, β-acetyl, γ-hexadecyl

  • CAS Number 99103-16-9

  • Empirical Formula (Hill Notation) C30H62NO7P

  • Molecular Weight 579.79

  •  MDL number MFCD00077384

  •  PubChem Substance ID 24895530

Purchase

Properties

Related Categories Cell Biology, Ether Lipid Metabolism, Metabolic Pathways, Metabolomics
InChI Key   HRYXIYMPLGFZGG-UHFFFAOYSA-N
assay   ~95%
storage temp.   −20°C

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?