• USA Home
  • H8774 - Bio-transport carrier system, Nalgene®
EMAIL THIS PAGE TO A FRIEND
H8774 Sigma
Bio-transport carrier system, Nalgene®
Carrier handle
Purchase
Properties
Related Categories Bottle Accessories, Bottles and Caps, Labware, Manufacturer Browser, Nalgene,
material   stainless steel
mfr. no.   Nalgene
Description
General description
Transparent, autoclavable system for safety transporting potentially biohazardous samples. Carrier is polycarbonate with a leakproof silicone gasket and polysulfone clamps. Handle is stainless steel.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?