• USA Home
  • H8884 - DL-S-β-Hydroxybutyryl-N-acetylcysteamine

EMAIL THIS PAGE TO A FRIEND
H8884 Sigma

DL-S-β-Hydroxybutyryl-N-acetylcysteamine

~98%

Purchase

Properties

assay   ~98%
storage temp.   −20°C

Description

Preparation Note

Chromatographically purified

Substrates

Substrate for the quantitative cytochemical determination of β-hydroxyacyl CoA dehydrogenase in heart muscle.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?