• USA Home
  • H9035 - Hemocyanin from mollusk

EMAIL THIS PAGE TO A FRIEND
H9035 Sigma

Hemocyanin from mollusk

from Concholepas sp., buffered aqueous glycerol solution

Synonym: Blue Carrier®

Purchase

Properties

Related Categories Antibodies, Antibody Purification and Characterization, Carrier Proteins, Supplementary Products
biological source   from Concholepas sp.
form   buffered aqueous glycerol solution
shipped in   wet ice
storage temp.   −20°C

Description

Biochem/physiol Actions

Highly soluble carrier protein used in antibody production

Physical form

Solution in 0.15 M NaCl, 0.05 M sodium phosphate, pH 7.2, and 50% glycerol.

Legal Information

Blue Carrier is a registered trademark of Biosonda S.A.

Price and Availability


Antibody Explorer

Explore with Confidence

Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?