EMAIL THIS PAGE TO A FRIEND
H9136 Sigma

HEPES

Synonym: N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) hemisodium salt

Purchase

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
TL6809000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

H4034 HEPES, BioPerformance Certified, ≥99.5% (titration), cell culture tested

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?