• USA Home
  • H9160 - Hydroxystilbamidine bis(methanesulfonate)

EMAIL THIS PAGE TO A FRIEND
H9160 Sigma

Hydroxystilbamidine bis(methanesulfonate)

Synonym: 2-Hydroxystilbene-4,4′-dicarboxamidine bis(methanesulfonate), Fluoro-Gold

  • CAS Number 223769-64-0

  • Empirical Formula (Hill Notation) C18H24N4O7S2

  • Molecular Weight 472.54

  •  MDL number MFCD01863083

Purchase

Properties

InChI Key   YGNSQKCULHSJDC-HFPMQDOPSA-N

Description

Legal Information

Fluoro-Gold is a trademark of Fluorochrome, Inc.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?