• USA Home
  • H9266 - Heparin sodium salt from sheep intestinal mucosa

EMAIL THIS PAGE TO A FRIEND
H9266 Sigma

Heparin sodium salt from sheep intestinal mucosa

~160 USP units/mg

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
MI0850000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

H3393 Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?